Please login to your online account to display your discounted pricing

Rat anti-B7-2/CD86, Clone: GL1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14338105MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1433815

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Mouse
Antigen B7-2/CD86
Formulation Phosphate buffer pH 7.2
Host Species Rat
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Immunogen The immunogen for this antibody was CD86.
Isotype IgG2a, κ
Purification Method Affinity Purified
Quantity 0.5mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 942
Concentration 0.5mg/mL
Format Affinity Purified
Clone GL1
Conjugate Unlabeled