Please login to your online account to display your discounted pricing

Rat anti-B7-2/CD86, Clone: GL1, FITC, Novus Biologicals™

$171.00 - $493.00

Specifications

Antigen B7-2/CD86
Clone GL1
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD86.
Gene ID (Entrez) 942
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14398801

 
novus biologicals canada
NBP14398801MG
0.1mg Each for $246.00

NBP14398805

 
novus biologicals canada
NBP143988005MG
0.05mg Each for $171.00

NBP1439885

 
novus biologicals canada
NBP14398805MG
0.5mg Each for $493.00
Description & Specifications

Specifications

Antigen B7-2/CD86
Clone GL1
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD86.
Gene ID (Entrez) 942
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry