Please login to your online account to display your discounted pricing

Rat anti-B220/CD45R, Clone: RA3-6B2, Biotin, Novus Biologicals™

$131.00 - $329.00

Specifications

Antigen B220/CD45R
Clone RA3-6B2
Concentration 0.5mg/mL
Immunogen Abelson murine leukemia virus-induced pre-B tumor cells
Gene ID (Entrez) 5788
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14364301

 
novus biologicals canada
NBP14364301MG
0.1mg Each for $197.00

NBP14364305

 
novus biologicals canada
NBP143643005MG
0.05mg Each for $131.00

NBP1436435

 
novus biologicals canada
NBP14364305MG
0.5mg Each for $329.00
Description & Specifications

Specifications

Antigen B220/CD45R
Clone RA3-6B2
Concentration 0.5mg/mL
Immunogen Abelson murine leukemia virus-induced pre-B tumor cells
Gene ID (Entrez) 5788
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry