Please login to your online account to display your discounted pricing

Rat anti-4-1BB Ligand/TNFSF9, Clone: TKS-1, PE, Novus Biologicals™

$289.00 - $418.00

Specifications

Antigen 4-1BB Ligand/TNFSF9
Clone TKS-1
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody is TNFRSF9.
Gene ID (Entrez) 8744
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20036010

 
novus biologicals canada
NBP200360100UG
100μg Each for $289.00

NBP20036026

 
novus biologicals canada
NBP200360200UG
200μg Each for $418.00
Description & Specifications

Specifications

Antigen 4-1BB Ligand/TNFSF9
Clone TKS-1
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody is TNFRSF9.
Gene ID (Entrez) 8744
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry