Please login to your online account to display your discounted pricing

Rat anti-2B4/CD244/SLAMF4, Clone: 244F4, Novus Biologicals™

$285.00 - $406.00

Specifications

Antigen 2B4/CD244/SLAMF4
Clone 244F4
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody is CD244.
Gene ID (Entrez) 51744
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20022355

 
novus biologicals canada
NBP200223500UG
500μg Each for $406.00

NBP20022310

 
novus biologicals canada
NBP200223100UG
100μg Each for $285.00
Description & Specifications

Specifications

Antigen 2B4/CD244/SLAMF4
Clone 244F4
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody is CD244.
Gene ID (Entrez) 51744
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Western Blotting