Please login to your online account to display your discounted pricing

Rat anti-2B4/CD244/SLAMF4, Clone: 244F4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200223500UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20022355

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Western Blotting
Gene ID (Entrez) 51744
Quantity 500μg
Cross Reactivity Mouse
Primary or Secondary Primary
Format Affinity Purified
Conjugate Unlabeled
Purification Method Affinity Purified
Immunogen The immunogen for this antibody is CD244.
Antigen 2B4/CD244/SLAMF4
Isotype IgG2a
Host Species Rat
Storage Requirements Store at 4°C. Do not freeze.
Regulatory Status RUO
Clone 244F4
Concentration 0.5mg/mL
Monoclonal or Polyclonal Monoclonal