Please login to your online account to display your discounted pricing

Rat anti-2B4/CD244/SLAMF4, Clone: 244F4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200223100UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20022310

  • / Each
 

Description & Specifications

Specifications

Antigen 2B4/CD244/SLAMF4
Applications Flow Cytometry
Applications Western Blot
Clone 244F4
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Affinity Purified
Host Species Rat
Immunogen The immunogen for this antibody is CD244.
Isotype IgG2a
Purification Method Affinity Purified
Quantity 100μg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 51744
Target Species Murine