missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

RASSF1A Mouse anti-Human, Clone: eB114-10H1, eBioscience™

Mouse Monoclonal Antibody

$130.68 - $328.68

Specifications

Antigen RASSF1A
Clone eB114-10H1
Host Species Murine
Species Reactivity Human
Applications Immunoprecipitation,Western Blot
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013558 Life Technologies
LS14688880
25μg Each for $130.68
5013559 Life Technologies
LS14688882
100μg Each for $328.68
Specifications

Description

The eB114 antibody reacts with the RASSF1A isoform of the tumor suppressor gene RASSF1. RASSF1A contains a Ras Association Domain (RA) and binds Ras in a GTP-dependent manner to effect apoptosis. The RASSF1A isoform undergoes epigenetic inactivation in a variety of human primary tumors. This has been associated with hypermethylation of a CpG island(s) in its promoter region. This antibody was generated against a synthetic peptide corresponding to the splice junction region of RASSF1A.
Description & Specifications

Specifications

RASSF1A
Murine
Immunoprecipitation,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
eB114-10H1
Human
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer