MP Biomedicals™ RapidTaq DNA Polymerase
RapidTaq DNA Polymerase is a next generation Taq polymerase for rapid DNA amplification in PCR reactions.
Manufacturer:  MP Biomedicals™ 116187200
Catalog No. MP116187200
Description
Description
RapidTaq is a unique DNA polymerase that is formulated for rapid DNA amplification in PCR reactions. With unique PCR reaction conditions, reaction times are reduced by up to 70% compared to wild-type Taq polymerase. PCR reaction time for standard templates under 1KB is reduced to ∼ 25 minutes with an increase of 7.5 minutes for every additional kb of amplified template using 30 cycles
  • Increased sensitivity, yields, and processivity
  • Rapid extension rates and robust performance
  • Products amplified with RapidTaq are compatible with TA cloning vectors
  • qPCR products amplified using the RapidTaq DNA polymerase can be used downstream for TA cloning
  • Primers and dNTPs not supplied
Specifications
Specifications
Polymerase + Buffer
-20°C
RapidTaq DNA polymerase is a thermostable DNA polymerase that is used for the DNA polymerase chain reaction (PCR) in order to amplify DNA sequences
DNA Polymerase (200 μL, 5X Taq Buffer (2 x 1.0 mL)
RapidTaq (5'-3' polymerase)
400
5’-3’ exonuclease activity, lacks 3’-5’ exonuclease activity with 3’-dA-tailed amplicons
SDS
Documents
Product Certifications
Promotions
Promotions
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.