Please login to your online account to display your discounted pricing

Rapid Decalcifier Solution, OSTEOMOLL™, EMD Millipore™

$151.91 - $337.09

Chemical Identifiers

Chemical Name or Material Rapid Decalcifier Solution
CAS 7647-01-0
CAS 50-00-0
CAS 67-56-1
UN Number UN 1789
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

M1017361000

 
emd millipore corporation
1017361000
Each for $151.91

M1017362500

 
emd millipore corporation
1017362500
Each for $337.09
Description & Specifications

Specifications

DOT Information DOT Class: 8, Packaging Group: II
CAS 7647-01-0
CAS 67-56-1
Packaging Plastic bottle
Chemical Name or Material Rapid Decalcifier Solution
Odor Stinging
Health Hazard 1 H290: May be corrosive to metals.
H302: Harmful if swallowed.
H315: Causes skin irritation.
H317: May cause an allergic skin reaction.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
H351: Suspected of causing cancer.
UN Number UN 1789
CAS 50-00-0
Appearance Blue liquid
Physical Form Liquid

For histology

Safety and Handling

H290: May be corrosive to metals.
H302: Harmful if swallowed.
H315: Causes skin irritation.
H317: May cause an allergic skin reaction.
H319: Causes serious eye irritation.
H335: May cause respiratory irritation.
H351: Suspected of causing cancer.