missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ranitidine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America R007325G
Specifications
Ranitidine Hydrochloride | |
135°C | |
C13H23ClN4O3S | |
25 g | |
GGWBHVILAJZWKJ-CHHCPSLASA-N | |
(Z)-1-N'-[2-[[5-[(dimethylamino)methyl]furan-2-yl]methylsulfanyl]ethyl]-1-N-methyl-2-nitroethene-1,1-diamine;hydrochloride | |
6603542 | |
≥98.0% (HPLC,T) |
66357-59-3 | |
White-Yellow | |
MFCD00069339 | |
ranitidine hydrochloride, noctone, n-2-5-dimethylamino methyl-2-furanyl methyl thio ethyl-n'-methyl-2-nitro-1,1-ethanediamine hydrochloride, c13h22n4o3s.hcl, opera_id_624, melfax hydrochloride, zantac hydrochloride, zintac hydrochloride, azantac hydrochloride, raniben hydrochloride | |
CNC(=C[N+](=O)[O-])NCCSCC1=CC=C(O1)CN(C)C.Cl | |
350.862 | |
350.86 | |
Crystalline Powder |
Chemical Identifiers
66357-59-3 | |
350.862 | |
GGWBHVILAJZWKJ-CHHCPSLASA-N | |
6603542 | |
CNC(=C[N+](=O)[O-])NCCSCC1=CC=C(O1)CN(C)C.Cl |
C13H23ClN4O3S | |
MFCD00069339 | |
ranitidine hydrochloride, noctone, n-2-5-dimethylamino methyl-2-furanyl methyl thio ethyl-n'-methyl-2-nitro-1,1-ethanediamine hydrochloride, c13h22n4o3s.hcl, opera_id_624, melfax hydrochloride, zantac hydrochloride, zintac hydrochloride, azantac hydrochloride, raniben hydrochloride | |
(Z)-1-N'-[2-[[5-[(dimethylamino)methyl]furan-2-yl]methylsulfanyl]ethyl]-1-N-methyl-2-nitroethene-1,1-diamine;hydrochloride |
Safety and Handling
RTECSNumber : KM6557000
TSCA : No
Recommended Storage : Refrigerator