Please login to your online account to display your discounted pricing

Ralfinamide mesylate, Tocris™

Na+ channel blocker

$205.70 - $862.95

Chemical Identifiers

Chemical Name or Material Ralfinamide mesylate
Assay Percent Range >99%
Molecular Formula C17H19FN2O2·CH3SO3H
Formula Weight 398.45
Synonym (2S)-2-[[[4-[(2-Fluorophenyl)methoxy]phenyl]methyl]amino]-propanamide methanesulfonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

402910

 
r & d systems
RDS402910
10mg Each for $205.70

402950

 
r & d systems
RDS402950
50mg Each for $862.95
Description & Specifications

Specifications

Formula Weight 398.45
Quantity 10mg
Chemical Name or Material Ralfinamide mesylate
Assay Percent Range >99%
Molecular Formula C17H19FN2O2·CH3SO3H
CAS 202825-45-4
Synonym (2S)-2-[[[4-[(2-Fluorophenyl)methoxy]phenyl]methyl]amino]-propanamide methanesulfonate
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C