missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ RAGE antagonist peptide
Description
RAGE antagonist peptide is a receptor for advanced glycation end products (RAGE) antagonist. Blocks S100P, S100A4 and HMGB-1 mediated RAGE activation in vitro and in vivo. Inhibits growth and metastasis of rat glioma tumors. Reduces cell growth and RAGE-mediated NF-κB activity in human PDAC cell lines. Inhibits effects of TDI exposure in BALB/c mice.
Specifications
Specifications
| Species | Human |
| Purity | 95% |
| CAS | 1092460-91-7 |
| Content And Storage | Store at −20°C |
| Molecular Weight (g/mol) | 1272.56g/mol |
| Quantity | 1 mg |
| Regulatory Status | RUO |
| Sequence | ELKVLMEKEL |
For Research Use Only