Alfa Aesar™ RAF265
Manufacturer:  Alfa Aesar™ J67037LB0
 View more versions of this product
Catalog No. AAJ67037LB0
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications
Specifications
Solid
5mg
Pale yellow to yellow
927880-90-8
-30°C to -10°C
C24H16F6N6O
RAF265
518.41
CHIR-265;
Light Sensitive