Please login to your online account to display your discounted pricing

Alfa Aesar™ RAF265


Manufacturer: alfa aesar™  J67037MA

 View more versions of this product

Catalog No. AAJ67037MA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Formula Weight 518.41
Description Pale yellow to yellow
Quantity 10mg
Storage Requirements -30°C to -10°C
Format Solid
Type RAF265
CAS 927880-90-8
Sensitivity Light Sensitive
Molecular Formula C24H16F6N6O
Synonym CHIR-265;