Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Raf-1 (Phospho-specific) Ab-2, Rat Monoclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ Raf-1 (Phospho-specific) Ab-2, Rat Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RT1732PABX

 View more versions of this product

Catalog No. RT1732PABX

  • / Each
 

Description & Specifications

Specifications

Antigen Raf-1 (Phospho-specific) Ab-2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone pS338[DPR/2/2A/3A]
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Formulation Ab without BSA/Azide (Frozen)
Host Species Rat
Immunogen A synthetic peptide corresponding to aa GQRDSasteriskSYYasteriskWEIEAS
Isotype IgG1
Quantity 200μL
Regulatory Status RUO
Research Discipline Signal Transduction
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human

Provide accurate, reproducible results with the Thermo Scientific™ Raf-1 (Phospho-specific) Ab-2, Rat Monoclonal Antibody.

Raf1 is an important mediator of signal transduction pathway. It is activated through activation by Src and protein kinase C. It further phosphorylates Mek1 and cdc25. It can bind to GTP-bound Ras through N-terminal domain. It also interacts with hsp90 and 14.3.3 family of proteins.

Host Species: Rat

Clone: pS338[DPR/2/2A/3A]

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: GQRDS™SYY™WEIEAS

Immunogen: A synthetic peptide corresponding to aa GQRDS™SYY™WEIEAS

Molecular Weight: 74kDa

Positive Control: Tonsil

Cellular Localization: Nuclear Ab-2 binds to phosphorylated serine residue at aa338. It does not bind to unactivated Raf-1 or Raf-1 phosphorylated at Y341 and S339.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO