Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Raf-1 (Phospho-specific) Ab-2, Rat Monoclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ Raf-1 (Phospho-specific) Ab-2, Rat Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RT1732P1ABX

 View more versions of this product

Catalog No. RT1732P1ABX

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry (Paraffin)
Antigen Raf-1 (Phospho-specific) Ab-2
Host Species Rat
Isotype IgG1
Format Concentrated
Immunogen A synthetic peptide corresponding to aa GQRDSasteriskSYYasteriskWEIEAS
Clone pS338[DPR/2/2A/3A]
Cross Reactivity Human
Quantity 100µL
Regulatory Status RUO
Formulation Ab without BSA/Azide (Frozen)
Conjugate Unlabeled
Research Discipline Signal Transduction
Type Primary
Classification Monoclonal
Concentration 1mg/mL
Item Description 100uL; 1mg/mL; Purified w/o BSA, azide

Provide accurate, reproducible results with the Thermo Scientific™ Raf-1 (Phospho-specific) Ab-2, Rat Monoclonal Antibody.

Raf1 is an important mediator of signal transduction pathway. It is activated through activation by Src and protein kinase C. It further phosphorylates Mek1 and cdc25. It can bind to GTP-bound Ras through N-terminal domain. It also interacts with hsp90 and 14.3.3 family of proteins.

Host Species: Rat

Clone: pS338[DPR/2/2A/3A]

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: GQRDS™SYY™WEIEAS

Immunogen: A synthetic peptide corresponding to aa GQRDS™SYY™WEIEAS

Molecular Weight: 74kDa

Positive Control: Tonsil

Cellular Localization: Nuclear Ab-2 binds to phosphorylated serine residue at aa338. It does not bind to unactivated Raf-1 or Raf-1 phosphorylated at Y341 and S339.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO