missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

RAE1 delta Mouse anti-Mouse, Clone: RD-41, eBioscience™

Mouse Monoclonal Antibody

$91.08 - $143.55

Specifications

Antigen RAE1 delta
Clone RD-41
Host Species Murine
Gene Alias RAE-1 delta, RAE1d
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013130 Life Technologies
LS14575681
50μg Each for $91.08
5013131 Life Technologies
LS14575682
100μg Each for $143.55
Specifications

Description

The RD-41 monoclonal antibody reacts with the murine Rae1δ. Rae-1δ is one of several known murine NKG2D ligands that include RAE-1 molecules (α, β, ε, γ and δ), H60 and MULT-1. Expression of NKG2D ligands is low or absent on normal adult tissues. However, stressed or transformed cells express NKG2D ligands which in turn activates NK cells tumoricidal activity through NKG2D. Until now, the expression of NKG2D ligands has been mainly studied with NKG2D tetramers which recognizes all NKG2D ligands. The RD-41 antibody has been reported to block tetramer staining.
Description & Specifications

Specifications

RAE1 delta
Murine
Mouse
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
RD-41
RAE-1 delta, RAE1d
ELISA,Flow Cytometry,Functional Assay,Immunohistochemistry,Immunohistochemistry (Frozen)
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL