Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ RAD51 Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ RAD51 Ab-1, Mouse Monoclonal Antibody.

$310.39 - $1,622.33

Specifications

Antigen RAD51 Ab-1
Clone 51RAD01; same as 3C10
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS988B

 
thermo scientific™ lab vision™
MS988B
Biotinylated Ab with BSA and Azide 1mL Each for $1,622.33

MS988B0

 
thermo scientific™ lab vision™
MS988B0
Biotinylated Ab with BSA and Azide 100μL Each for $405.86

MS988B1

 
thermo scientific™ lab vision™
MS988B1
Biotinylated Ab with BSA and Azide 500μL Each for $1,044.51

MS988P

 
thermo scientific™ lab vision™
MS988P
Purified Ab with BSA and Azide 1mL Each for $1,292.35

MS988P0

 
thermo scientific™ lab vision™
MS988P0
Purified Ab with BSA and Azide 100μL Each for $310.39

MS988P1

 
thermo scientific™ lab vision™
MS988P1
Purified Ab with BSA and Azide 500μL Each for $815.71

MS988P1ABX

 
thermo scientific™ lab vision™
MS988P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $879.39

MS988PABX

 
thermo scientific™ lab vision™
MS988PABX
Ab without BSA/Azide (Frozen) 200μL Each for $1,413.02

MS988R7

 
thermo scientific™ lab vision™
MS988R7
Ready-to-use for IHC (UVLP) 7mL Each for $835.20
Description & Specifications

Specifications

Antigen RAD51 Ab-1
Clone 51RAD01; same as 3C10
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Host Species Murine
Regulatory Status RUO
Research Discipline DNA Repair
Type Primary
Immunogen Recombinant Rad51 protein
Isotype IgG2a kappa

Provide accurate, reproducible results with the Thermo Scientific™ RAD51 Ab-1, Mouse Monoclonal Antibody.

Rad51 (a eukaryotic homologue of Escherichia coli RecA) is similar to RecA, both biochemically and structurally: It promotes homologous pairing and strand exchange within a regular nucleoprotein filament. In yeast, the Rad51 family of related proteins also includes Rad55, Rad57 and Dmc1. In mammalian cells, seven genes in this family have been identified (HsRAD51, XRCC2, XRCC3, RAD51B/hREC2 and HsDMC1, RAD51C, and RAD 51D). Recent studies suggested that Rad51 is involved not only in homologous recombination but also in cell proliferation regulation.

Host Species: Mouse

Clone: 51RAD01; same as 3C10

Isotype: IgG1/κ

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: Not determined

Immunogen: Recombinant RAD51 protein

Molecular Weight: 37-43kDa

Positive Control: HeLa cells, Testis

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO