Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ RAD51 Ab-1, Mouse Monoclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ RAD51 Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS988P1ABX

 View more versions of this product

Catalog No. MS988P1ABX

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Regulatory Status RUO
Clone 51RAD01; same as 3C10
Antigen RAD51 Ab-1
Immunogen Recombinant Rad51 protein
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Format Concentrated
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgG2a kappa
Quantity 100µL
Type Primary
Concentration 1mg/mL
Research Discipline DNA Repair
Classification Monoclonal
Conjugate Unlabeled
Formulation Ab without BSA/Azide (Frozen)

Provide accurate, reproducible results with the Thermo Scientific™ RAD51 Ab-1, Mouse Monoclonal Antibody.

Rad51 (a eukaryotic homologue of Escherichia coli RecA) is similar to RecA, both biochemically and structurally: It promotes homologous pairing and strand exchange within a regular nucleoprotein filament. In yeast, the Rad51 family of related proteins also includes Rad55, Rad57 and Dmc1. In mammalian cells, seven genes in this family have been identified (HsRAD51, XRCC2, XRCC3, RAD51B/hREC2 and HsDMC1, RAD51C, and RAD 51D). Recent studies suggested that Rad51 is involved not only in homologous recombination but also in cell proliferation regulation.

Host Species: Mouse

Clone: 51RAD01; same as 3C10

Isotype: IgG1/κ

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: Not determined

Immunogen: Recombinant RAD51 protein

Molecular Weight: 37-43kDa

Positive Control: HeLa cells, Testis

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO