missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Raclopride 98.0+%, TCI America™

Manufacturer:  Tci America R0094200MG

 View more versions of this product

Catalog No. R0094200MG



Specifications

Specifications

Raclopride
54°C
84225-95-6
MFCD02179386
WAOQONBSWFLFPE-VIFPVBQESA-N
3,5-dichloro-N-[[(2S)-1-ethylpyrrolidin-2-yl]methyl]-2-hydroxy-6-methoxybenzamide
3033769
Crystal-Powder at 20°C
Undesignated
200mg
C15H20Cl2N2O3
raclopride, s-3,5-dichloro-n-1-ethylpyrrolidin-2-yl methyl-2-hydroxy-6-methoxybenzamide, 3h raclopride, 3h-raclopride, unii-430k3soz7g, chembl8809, raclopridum latin, racloprida spanish, raclopride inn:ban, dsstox_cid_25687
CCN1CCCC1CNC(=O)C2=C(C(=CC(=C2OC)Cl)Cl)O
347.236
347.24
≥98.0% (HPLC,N)

Chemical Identifiers

84225-95-6
347.236
WAOQONBSWFLFPE-VIFPVBQESA-N
3033769
CCN1CCCC1CNC(=O)C2=C(C(=CC(=C2OC)Cl)Cl)O
C15H20Cl2N2O3
MFCD02179386
raclopride, s-3,5-dichloro-n-1-ethylpyrrolidin-2-yl methyl-2-hydroxy-6-methoxybenzamide, 3h raclopride, 3h-raclopride, unii-430k3soz7g, chembl8809, raclopridum latin, racloprida spanish, raclopride inn:ban, dsstox_cid_25687
3,5-dichloro-N-[[(2S)-1-ethylpyrrolidin-2-yl]methyl]-2-hydroxy-6-methoxybenzamide
Safety and Handling

Safety and Handling

RTECSNumber : CV3463500

TSCA : No

Documents