Please login to your online account to display your discounted pricing

rac-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl, +96%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC392230010

 View more versions of this product

Catalog No. AC392230010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 678.79
Physical Form Crystalline Powder
Quantity 1g
Color White to off-white
Molecular Formula C48H40P2
Chemical Name or Material rac-2, 2'-Bis(di-p-tolylphosphino)-1, 1'-binaphthyl
CAS 153305-67-0
MDL Number MFCD01311709
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 96+%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material rac-2, 2'-Bis(di-p-tolylphosphino)-1, 1'-binaphthyl
CAS 153305-67-0
Assay ≥96.0%
Assay Percent Range 96+%
Molecular Formula C48H40P2
Formula Weight 678.79
MDL Number MFCD01311709
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents