Please login to your online account to display your discounted pricing

rac-2,2'-Bis(di-p-tolylphosphino)-1,1'-binaphthyl, +96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material rac-2, 2'-Bis(di-p-tolylphosphino)-1, 1'-binaphthyl
CAS 153305-67-0
Assay ≥96.0%
Assay Percent Range 96+%
Molecular Formula C48H40P2
Formula Weight 678.79
MDL Number MFCD01311709
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC392230050

  SDS
thermo fisher scientific
AC392230050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC392230010

  SDS
thermo fisher scientific
AC392230010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 678.79
Physical Form Crystalline Powder
Quantity 5g
Color White to off-white
Packaging Glass bottle
CAS 153305-67-0
Chemical Name or Material rac-2, 2'-Bis(di-p-tolylphosphino)-1, 1'-binaphthyl
MDL Number MFCD01311709
Assay Percent Range 96+%
Infrared Spectrum Authentic
Molecular Formula C48H40P2
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.