Please login to your online account to display your discounted pricing

rac-2,2'-Bis(di(3,5-dimethylphenyl)phosphino)-1,1'-binaphthyl, +97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material rac-2,2'-Bis(di(3,5-dimethylphenyl)phosphino)-1,1'-binaphthyl
CAS 145416-77-9
Assay 97.0% min.
Assay Percent Range 97+%
Molecular Formula C52H48P2
Formula Weight 734.9g/mol
MDL Number MFCD01630821
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC392220010

  SDS
thermo fisher scientific
392220010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC392222500

  SDS
thermo fisher scientific
392222500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Color Pale yellow
Quantity 1g
Chemical Name or Material rac-2,2'-Bis(di(3,5-dimethylphenyl)phosphino)-1,1'-binaphthyl
CAS 145416-77-9
Physical Form Pale yellow crystalline powder
Assay 97.0% min.
Assay Percent Range 97+%
Molecular Formula C52H48P2
Formula Weight 734.9g/mol
MDL Number MFCD01630821
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.