Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Rabies Virus Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Rabies Virus Ab-1, Mouse Monoclonal Antibody.

$236.71 - $1,191.88

Specifications

Antigen Rabies Virus Ab-1
Clone Rab-50
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Rabies Virus
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS549P

 
thermo scientific™ lab vision™
MS549P
200μg/mL 1mL Each for $1,109.04

MS549P0

 
thermo scientific™ lab vision™
MS549P0
200μg/mL 100μL Each for $236.71

MS549P1

 
thermo scientific™ lab vision™
MS549P1
200μg/mL 500μL Each for $695.96

MS549P1ABX

 
thermo scientific™ lab vision™
MS549P1ABX
1mg/mL 100μL Each for $788.80

MS549PABX

 
thermo scientific™ lab vision™
MS549PABX
1mg/mL 200μL Each for $1,191.88
Description & Specifications

Specifications

Antigen Rabies Virus Ab-1
Clone Rab-50
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Rabies Virus
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline Infectious Disease
Type Primary
Immunogen Rabies Virus of strains SAD-Vnukovo and Pitman Moore
Isotype IgG2b kappa

Provide accurate, reproducible results with the Thermo Scientific™ Rabies Virus Ab-1, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: Rab-50

Isotype: IgG2b/κ

Species Reactivity: Rabies virus of strains SAD-Vnukovo and Pitman Moore

Epitope: Not determined

Immunogen: Rabies Virus of strains SAD-Vnukovo and Pitman Moore Rab-50 antibody reacts with viral glycoprotein of rabies virus strains SAD-Vnukovo and Pitman-Moore. It can be used for detecting rabies virus by ELISA and Western. It is capable of neutralizing rabies virus.

Recommended for:

  • ELISA
  • Neutralization in vitro

Note:

USA: RUO; Int'l: RUO