Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Rabies Virus Ab-1, Mouse Monoclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ Rabies Virus Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS549P1ABX

 View more versions of this product

Catalog No. MS549P1ABX

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Cross Reactivity Rabies Virus
Quantity 100µL
Regulatory Status RUO
Antigen Rabies Virus Ab-1
Immunogen Rabies Virus of strains SAD-Vnukovo and Pitman Moore
Isotype IgG2b kappa
Clone Rab-50
Format Concentrated
Type Primary
Classification Monoclonal
Conjugate Unlabeled
Research Discipline Infectious Disease
Concentration 1mg/mL

Provide accurate, reproducible results with the Thermo Scientific™ Rabies Virus Ab-1, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: Rab-50

Isotype: IgG2b/κ

Species Reactivity: Rabies virus of strains SAD-Vnukovo and Pitman Moore

Epitope: Not determined

Immunogen: Rabies Virus of strains SAD-Vnukovo and Pitman Moore Rab-50 antibody reacts with viral glycoprotein of rabies virus strains SAD-Vnukovo and Pitman-Moore. It can be used for detecting rabies virus by ELISA and Western. It is capable of neutralizing rabies virus.

Recommended for:

  • ELISA
  • Neutralization in vitro

Note:

USA: RUO; Int'l: RUO