missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabeprazole Sulfide 98.0+%, TCI America™
Supplier: TCI America R00981G
Specifications
| Rabeprazole Sulfide | |
| 116°C | |
| C18H21N3O2S | |
| 1 g | |
| BSXAHDOWMOSVAP-UHFFFAOYSA-N | |
| 2-({[4-(3-methoxypropoxy)-3-methylpyridin-2-yl]methyl}sulfanyl)-1H-1,3-benzodiazole | |
| 9949996 | |
| ≥98.0% (HPLC,T) |
| 117977-21-6 | |
| White-Yellow | |
| MFCD08063845 | |
| 2-[[4-(3-Methoxypropoxy)-3-methyl-2-pyridyl]methylthio]benzimidazole | |
| COCCCOC1=C(C)C(CSC2=NC3=CC=CC=C3N2)=NC=C1 | |
| 343.45 | |
| 343.45 | |
| Crystalline Powder |
Chemical Identifiers
| 117977-21-6 | |
| 343.45 | |
| BSXAHDOWMOSVAP-UHFFFAOYSA-N | |
| 9949996 | |
| COCCCOC1=C(C)C(CSC2=NC3=CC=CC=C3N2)=NC=C1 |
| C18H21N3O2S | |
| MFCD08063845 | |
| 2-[[4-(3-Methoxypropoxy)-3-methyl-2-pyridyl]methylthio]benzimidazole | |
| 2-({[4-(3-methoxypropoxy)-3-methylpyridin-2-yl]methyl}sulfanyl)-1H-1,3-benzodiazole |
Safety and Handling
TSCA : No