missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Rabeprazole Sodium Salt 98.0+%, TCI America™
Supplier: TCI America R0115100MG
Specifications
Rabeprazole Sodium Salt | |
150°C | |
C18H20N3NaO3S | |
100 mg | |
[Na+].COCCCOC1=CC=NC(CS(=O)C2=NC3=CC=CC=C3[N-]2)=C1C | |
381.43 | |
CHEBI:8769 | |
≥98.0% (HPLC) |
117976-90-6 | |
White-Yellow | |
MFCD02092688 | |
KRCQSTCYZUOBHN-UHFFFAOYNA-N | |
sodium 2-{[4-(3-methoxypropoxy)-3-methylpyridin-2-yl]methanesulfinyl}-1H-1,3-benzodiazol-1-ide | |
14720269 | |
381.43 | |
Crystalline Powder |
Chemical Identifiers
117976-90-6 | |
381.43 | |
KRCQSTCYZUOBHN-UHFFFAOYNA-N | |
CHEBI:8769 | |
[Na+].COCCCOC1=CC=NC(CS(=O)C2=NC3=CC=CC=C3[N-]2)=C1C |
C18H20N3NaO3S | |
MFCD02092688 | |
14720269 | |
sodium 2-{[4-(3-methoxypropoxy)-3-methylpyridin-2-yl]methanesulfinyl}-1H-1,3-benzodiazol-1-ide |
Safety and Handling
TSCA : No
Recommended Storage : Freezer