missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Normal Rabbit Serum, Invitrogen™
Rabbit Control
Supplier: Invitrogen 016101
Description
Normal Rabbit Serum Control, 01-6101, from Invitrogen™. Species Reactivity: Not Applicable; Applications: Control.
Normal Sera are non-immune serum samples from goat, human, mouse, rabbit and other species that contain the usual complement of serum proteins, including the various classes of immunoglobulins that exist in healthy animals of the particular species. Normal serum provides a sufficient quantity of endogenous proteins to saturate and block nonspecific binding sites on fixed samples or assay substrates. When used as an antibody diluent, non-immune serum also supplies a native chemical environment for antibodies to function properly without adding foreign protein components. Normal serum is frequently used for blocking or saturating generalized binding interactions for immunodetection methods, especially those involving tissue samples such as immunohistochemistry (IHC). Normal sera are also useful as controls for testing general and specific antibody purification methods.Host Species: Rabbit
Isotype: Ig Mix
Immunogen: Rabbit immunoglobins
Formulation: 10mL unpurified control antibody in whole serum and 0.1% sodium azide
Reconstitution with 10mL of distilled water. After reconstitution, store product in frozen aliquots.
Order Info
Shipping Condition: Wet Ice
Specifications
Normal Rabbit Serum | |
whole serum with 0.1% sodium azide | |
Rabbit | |
10 mL | |
Isotype Control (Primary) | |
Lyophilized |
Unconjugated | |
Whole serum | |
Rabbit immunoglobins | |
RUO | |
None | |
Ig Mix |
For Research Use Only. Not for use in diagnostic procedures.