Please login to your online account to display your discounted pricing

Rabbit anti-Vinculin, Clone: SP117, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21250705ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21250705

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Cross Reactivity Human
Antigen Vinculin
Format Purified
Formulation PBS/1% BSA buffer pH 7.6
Host Species Rabbit
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Immunogen Synthetic peptide derived from C-terminus of human vinculin protein.
Isotype IgG
Purification Method Purified
Quantity 0.5mL
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7414
Test Specificity Predicted to cross react with Bovine, Chicken, Canine, Mouse, Porcine, Rat.
Reconstitution Protein A purified
Clone SP117
Conjugate Unlabeled