Please login to your online account to display your discounted pricing

Rabbit anti-Vinculin, Clone: SP117, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21250701ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21250701

  • / Each
 

Description & Specifications

Specifications

Antigen Vinculin
Formulation PBS/1% BSA buffer pH 7.6
Host Species Rabbit
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Immunogen Synthetic peptide derived from C-terminus of human vinculin protein.
Isotype IgG
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7414
Test Specificity Predicted to cross react with Bovine, Chicken, Canine, Mouse, Porcine, Rat.
Reconstitution Protein A purified
Clone SP117