Please login to your online account to display your discounted pricing

Rabbit anti-Villin 1, Clone: SP145, Novus Biologicals™

$309.05 - $551.93

Specifications

Antigen Villin 1
Clone SP145
Immunogen Synthetic peptide derived from the C-terminus of human villin protein.
Gene ID (Entrez) 7429
Formulation PBS/1% BSA buffer pH 7.6
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP21251305

 
novus biologicals canada
NBP21251305ML
0.5mL Each for $551.93

NBP21251301

 
novus biologicals canada
NBP21251301ML
0.1mL Each for $309.05
Description & Specifications

Specifications

Antigen Villin 1
Clone SP145
Immunogen Synthetic peptide derived from the C-terminus of human villin protein.
Gene ID (Entrez) 7429
Formulation PBS/1% BSA buffer pH 7.6
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Isotype IgG
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot