Please login to your online account to display your discounted pricing

Rabbit anti-Villin 1, Clone: SP145, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21251305ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21251305

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Storage Requirements Store at 4°C. Do not freeze.
Quantity 0.5mL
Isotype IgG
Formulation PBS/1% BSA buffer pH 7.6
Monoclonal or Polyclonal Monoclonal
Format Purified
Regulatory Status RUO
Test Specificity Predicted to cross react with Bovine, Chicken, Mouse, Porcine.
Primary or Secondary Primary
Conjugate Unlabeled
Host Species Rabbit
Antigen Villin 1
Immunogen Synthetic peptide derived from the C-terminus of human villin protein.
Gene ID (Entrez) 7429
Clone SP145
Purification Method Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Reconstitution Protein A purified