Please login to your online account to display your discounted pricing

Rabbit anti-Villin 1, Clone: SP145, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21251301ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21251301

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Isotype IgG
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Synthetic peptide derived from the C-terminus of human villin protein.
Quantity 0.1mL
Formulation PBS/1% BSA buffer pH 7.6
Antigen Villin 1
Purification Method Purified
Format Purified
Conjugate Unlabeled
Regulatory Status RUO
Cross Reactivity Human
Storage Requirements Store at 4°C. Do not freeze.
Clone SP145
Host Species Rabbit
Primary or Secondary Primary
Reconstitution Protein A purified
Gene ID (Entrez) 7429
Test Specificity Predicted to cross react with Bovine, Chicken, Mouse, Porcine.