Please login to your online account to display your discounted pricing

Rabbit anti-VEGF R2/KDR/Flk-1, Clone: SP123, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP17452005ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17452005

  • / Each
 

Description & Specifications

Specifications

Host Species Rabbit
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 3791
Conjugate Unlabeled
Cross Reactivity Human
Antigen VEGF R2/KDR/Flk-1
Formulation PBS 1% BSA buffer pH 7.6 with less than 0.1% sodium azide.
Immunogen Synthetic peptide near the N-terminus of mouse FLK-1 protein.
Isotype IgG
Format Purified
Purification Method Purified
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Clone SP123