Please login to your online account to display your discounted pricing

Rabbit anti-VEGF R2/KDR/Flk-1, Clone: SP123, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP17452010ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17452010

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Cross Reactivity Human
Antigen VEGF R2/KDR/Flk-1
Formulation PBS 1% BSA buffer pH 7.6 with less than 0.1% sodium azide.
Immunogen Synthetic peptide near the N-terminus of mouse FLK-1 protein.
Isotype IgG
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Format Purified
Host Species Rabbit
Purification Method Purified
Quantity 1mL
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 3791
Reconstitution Protein A purified
Clone SP123
Conjugate Unlabeled