Please login to your online account to display your discounted pricing

Rabbit anti-VEGF R2/KDR/Flk-1, Clone: SP123, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP17452001ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17452001

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Immunogen Synthetic peptide near the N-terminus of mouse FLK-1 protein.
Quantity 0.1mL
Formulation PBS 1% BSA buffer pH 7.6 with less than 0.1% sodium azide.
Isotype IgG
Conjugate Unlabeled
Clone SP123
Monoclonal or Polyclonal Monoclonal
Storage Requirements Store at 4°C. Do not freeze.
Antigen VEGF R2/KDR/Flk-1
Reconstitution Protein A purified
Purification Method Purified
Gene ID (Entrez) 3791
Host Species Rabbit
Format Purified
Regulatory Status RUO
Primary or Secondary Primary