Please login to your online account to display your discounted pricing

Rabbit anti-TTF1 / NKX2.1, Clone: G21-G, Novus Biologicals™

$367.83 - $569.00

Specifications

Antigen TTF1 / NKX2.1
Clone G21-G
Immunogen Peptide derived from N-terminal sequence of human TTF-1
Gene ID (Entrez) 7080
Formulation 20mM Tris-HCl, pH 8.0 20mg/mL BSA
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP17905610

 
novus biologicals canada
NBP179056100UL
100μL Each for $367.83

NBP17905620

 
novus biologicals canada
NBP179056200UL
200μL Each for $569.00
Description & Specifications

Specifications

Antigen TTF1 / NKX2.1
Clone G21-G
Immunogen Peptide derived from N-terminal sequence of human TTF-1
Gene ID (Entrez) 7080
Formulation 20mM Tris-HCl, pH 8.0 20mg/mL BSA
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Cross Reactivity Human
Isotype IgG
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)