Please login to your online account to display your discounted pricing

Rabbit anti-TTF1 / NKX2.1, Clone: G21-G, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP179056200UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17905620

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Monoclonal or Polyclonal Monoclonal
Isotype IgG
Host Species Rabbit
Primary or Secondary Primary
Conjugate Unlabeled
Immunogen Peptide derived from N-terminal sequence of human TTF-1
Quantity 200μL
Cross Reactivity Human
Format Affinity Purified
Clone G21-G
Antigen TTF1 / NKX2.1
Purification Method Affinity Purified
Formulation 20mM Tris-HCl, pH 8.0 20mg/mL BSA
Gene ID (Entrez) 7080
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.