Please login to your online account to display your discounted pricing

Rabbit anti-TTF1 / NKX2.1, Clone: C12-I, Novus Biologicals™

$397.00 - $569.00

Specifications

Antigen TTF1 / NKX2.1
Clone C12-I
Immunogen Peptide derived from C-terminal sequence of human TTF-1
Gene Accession No. P43699
Gene ID (Entrez) 7080
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP17905720

 
novus biologicals canada
NBP179057200UL
200μL Each for $569.00

NBP17905710

 
novus biologicals canada
NBP179057100UL
100μL Each for $397.00
Description & Specifications

Specifications

Antigen TTF1 / NKX2.1
Clone C12-I
Immunogen Peptide derived from C-terminal sequence of human TTF-1
Gene Accession No. P43699
Gene ID (Entrez) 7080
Formulation 20mM Tris-HCl, pH 8.0 20mg/mL BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Cross Reactivity Human
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)