Please login to your online account to display your discounted pricing

Rabbit anti-TTF1 / NKX2.1, Clone: C12-I, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP179057200UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17905720

  • / Each
 

Description & Specifications

Specifications

Antigen TTF1 / NKX2.1
Host Species Rabbit
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Storage Requirements Store at 4°C. Do not freeze.
Cross Reactivity Human
Format Affinity Purified
Formulation 20mM Tris-HCl, pH 8.0 20mg/mL BSA.
Gene Accession No. P43699
Immunogen Peptide derived from C-terminal sequence of human TTF-1
Quantity 200μL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone C12-I
Conjugate Unlabeled
Purification Method Affinity Purified
Gene ID (Entrez) 7080