Please login to your online account to display your discounted pricing

Rabbit anti-TTF1 / NKX2.1, Clone: C12-I, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP179057100UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17905710

  • / Each
 

Description & Specifications

Specifications

Antigen TTF1 / NKX2.1
Formulation 20mM Tris-HCl, pH 8.0 20mg/mL BSA.
Host Species Rabbit
Immunogen Peptide derived from C-terminal sequence of human TTF-1
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Format Affinity Purified
Gene Accession No. P43699
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Purification Method Affinity Purified
Quantity 100μL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Gene ID (Entrez) 7080
Clone C12-I
Conjugate Unlabeled