Please login to your online account to display your discounted pricing

Rabbit anti-Trypsin 1/PRSS1, Clone: A16-N, Novus Biologicals™

$364.17 - $364.17

Specifications

Antigen Trypsin 1/PRSS1
Clone A16-N
Concentration 2.7mg/mL
Immunogen Peptide derived from near the N terminus of human pancreatic cationic trypsin.
Gene Accession No. P07477
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP13013505

 
novus biologicals canada
NBP130135005ML
0.05mL Each for $364.17
Description & Specifications

Specifications

Antigen Trypsin 1/PRSS1
Clone A16-N
Concentration 2.7mg/mL
Immunogen Peptide derived from near the N terminus of human pancreatic cationic trypsin.
Gene Accession No. P07477
Gene ID (Entrez) 5644
Formulation 20mM Tris-HCl, pH 8.0 with 10mg/mL BSA. Immunogen affinity purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Rabbit
Isotype IgG
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications ELISA
Applications Western Blot