Please login to your online account to display your discounted pricing

Rabbit anti-Trypsin 1/PRSS1, Clone: A16-N, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13013501ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13013501

  • / Each
 

Description & Specifications

Specifications

Format Affinity Purified
Gene Accession No. P07477
Immunogen Peptide derived from near the N terminus of human pancreatic cationic trypsin.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 5644
Clone A16-N
Cross Reactivity Human
Antigen Trypsin 1/PRSS1
Formulation 20mM Tris-HCl, pH 8.0 with 10mg/mL BSA. Immunogen affinity purified.
Host Species Rabbit
Applications ELISA
Applications Western Blotting
Isotype IgG
Purification Method Affinity Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Test Specificity This antibody is specific for the N terminus of human pancreatic cationic trypsin.
Concentration 2.7mg/mL
Conjugate Unlabeled
Quantity 0.1mL