Please login to your online account to display your discounted pricing

Rabbit anti-Tensin 4/CTEN, Clone: SP83, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21248805ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21248805

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Quantity 0.5mL
Format Purified
Host Species Rabbit
Immunogen Synthetic peptide corresponding to C-terminus of human c-TEN protein.
Gene ID (Entrez) 84951
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Monoclonal or Polyclonal Monoclonal
Formulation PBS/1% BSA buffer pH 7.6
Conjugate Unlabeled
Test Specificity Predicted to cross react with Bovine, Mouse.
Clone SP83
Primary or Secondary Primary
Isotype IgG
Purification Method Purified
Antigen Tensin 4/CTEN
Storage Requirements Store at 4°C. Do not freeze.
Regulatory Status RUO
Reconstitution Protein A purified