Please login to your online account to display your discounted pricing

Rabbit anti-Tensin 4/CTEN, Clone: SP83, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21248801ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21248801

  • / Each
 

Description & Specifications

Specifications

Format Purified
Immunogen Synthetic peptide corresponding to C-terminus of human c-TEN protein.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Clone SP83
Isotype IgG
Regulatory Status RUO
Test Specificity Predicted to cross react with Bovine, Mouse.
Antigen Tensin 4/CTEN
Quantity 0.1mL
Gene ID (Entrez) 84951
Purification Method Purified
Formulation PBS/1% BSA buffer pH 7.6
Conjugate Unlabeled
Host Species Rabbit
Storage Requirements Store at 4°C. Do not freeze.
Reconstitution Protein A purified
Cross Reactivity Human