Please login to your online account to display your discounted pricing

Rabbit anti-Syntaxin 1A, Clone: SP36, Novus Biologicals™

$309.05 - $586.39

Specifications

Antigen Syntaxin 1A
Clone SP36
Immunogen Synthetic peptide corresponding to N-terminus of human Syntaxin 1A
Gene ID (Entrez) 6804
Formulation 10mM PBS, pH 7.4, with 1% BSA (7.0mL size Prediluted antibody). Immunogen affinity purified.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14980310

 
novus biologicals canada
NBP14980310ML
1mL Each for $586.39

NBP14980301

 
novus biologicals canada
NBP14980301ML
0.1mL Each for $309.05
Description & Specifications

Specifications

Antigen Syntaxin 1A
Clone SP36
Immunogen Synthetic peptide corresponding to N-terminus of human Syntaxin 1A
Gene ID (Entrez) 6804
Formulation 10mM PBS, pH 7.4, with 1% BSA (7.0mL size Prediluted antibody). Immunogen affinity purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Isotype IgG
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)