Please login to your online account to display your discounted pricing

Rabbit anti-Syntaxin 1A, Clone: SP36, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14980310ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14980310

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Antigen Syntaxin 1A
Monoclonal or Polyclonal Monoclonal
Formulation 10mM PBS, pH 7.4, with 1% BSA (7.0mL size Prediluted antibody). Immunogen affinity purified.
Cross Reactivity Human
Purification Method Affinity Purified
Quantity 1mL
Host Species Rabbit
Format Affinity Purified
Test Specificity May react with Mouse.
Primary or Secondary Primary
Isotype IgG
Gene ID (Entrez) 6804
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Immunogen Synthetic peptide corresponding to N-terminus of human Syntaxin 1A
Clone SP36