Please login to your online account to display your discounted pricing

Rabbit anti-Syntaxin 1A, Clone: SP36, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14980301ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14980301

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Gene ID (Entrez) 6804
Immunogen Synthetic peptide corresponding to N-terminus of human Syntaxin 1A
Cross Reactivity Human
Quantity 0.1mL
Monoclonal or Polyclonal Monoclonal
Storage Requirements Store at 4°C. Do not freeze.
Regulatory Status RUO
Clone SP36
Primary or Secondary Primary
Isotype IgG
Format Affinity Purified
Antigen Syntaxin 1A
Host Species Rabbit
Conjugate Unlabeled
Purification Method Affinity Purified
Test Specificity May react with Mouse.
Formulation 10mM PBS, pH 7.4, with 1% BSA (7.0mL size Prediluted antibody). Immunogen affinity purified.