Please login to your online account to display your discounted pricing

Rabbit anti-Slit2, Clone: SP122, Novus Biologicals™

$309.05 - $805.66

Specifications

Antigen Slit2
Clone SP122
Immunogen Synthetic peptide corresponding to C-terminus of human SLIT2 protein.
Gene ID (Entrez) 9353
Formulation PBS 1% BSA buffer pH 7.6 with less than 0.1% sodium azide.
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP17450401

 
novus biologicals canada
NBP17450401ML
0.1mL Each for $309.05

NBP17450405

 
novus biologicals canada
NBP17450405ML
0.5mL Each for $551.93

NBP17450410

 
novus biologicals canada
NBP17450410ML
1mL Each for $805.66
Description & Specifications

Specifications

Antigen Slit2
Clone SP122
Immunogen Synthetic peptide corresponding to C-terminus of human SLIT2 protein.
Gene ID (Entrez) 9353
Formulation PBS 1% BSA buffer pH 7.6 with less than 0.1% sodium azide.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Cross Reactivity Human
Cross Reactivity Rat
Isotype IgG
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)